missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Donkey anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP

Catalog No. SA1100
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA1100 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA1100 Supplier Invitrogen™ Supplier No. SA1100

Donkey Polyclonal Secondary Antibody

Product SA1-100 is conjugated to HRP and optimized for ECL applications. This antibody reacst with the heavy and light chains of the immunoglobulin molecule. Pre-absorbed with the following species proteins: Bovine, chicken, goat, guinea pig, syrian hamster, horse, human, rabbit, and sheep serum proteins.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Mouse IgG (H+L) Cross-Adsorbed
Applications Western Blot
Classification Polyclonal
Concentration 40 μg/mL
Conjugate HRP
Formulation PBS with 50% glycerol, proprietary stabilizer and 0.07% Kathon
Host Species Donkey
Purification Method Antigen affinity chromatography
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less