missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Mouse, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP192FMI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP192FMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP192FMI Supplier MilliporeSigma Supplier No. AP192F

Donkey Polyclonal Antibody

Detect Donkey Mouse IgG using this Donkey anti-Mouse IgG Antibody, FITC conjugate, Species Adsorbed validated for use in immunofluoresence.

Specifications

Antigen Donkey anti-Mouse IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation Lyophilized. Buffer = 0.02M Sodium Phosphate, 0.25M NaCl, pH 7.6, with 15mg/mL BSA and 0.1% sodium azide. RECONSTITUTION: Reconstitute to 1mg/mL with sterile distilled water. F/P RATIO: 15 micrograms FITC per mg of antibody.
Host Species Donkey
Purification Method Affinity Purified
Quantity 500 μg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only