missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Mouse, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP192CMI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP192CMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP192CMI Supplier MilliporeSigma Supplier No. AP192C

Donkey Polyclonal Antibody

This Donkey anti-Mouse IgG Antibody, Cy3 conjugate, Species Adsorbed is validated for use in IF for the detection of Donkey Mouse IgG.

Specifications

Antigen Donkey anti-Mouse IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Conjugate Cyanine3
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, and 0.05% sodium azide. RECONSTITUTION: Reconstitute with 500μL of sterile distilled water.
Host Species Donkey
Purification Method Affinity Purified
Quantity 500 μg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C in dark. Do not freeze.
Show More Show Less

For Research Use Only