missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Mouse, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP192BMI
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP192BMI 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP192BMI Supplier MilliporeSigma Supplier No. AP192B

Donkey Polyclonal Antibody

Donkey anti-Mouse IgG Antibody, biotin conjugate, Species Adsorbed detects level of Donkey Mouse IgG and has been published and validated for use in ELISA and western blotting.

Specifications

Antigen Donkey anti-Mouse IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.8mg/mL
Conjugate Biotin
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA and 0.01% thimerosal. RECONSTITUTION: Reconstitute with 500μL of sterile distilled water.
Host Species Donkey
Purification Method Immunoaffinity Purified and Immunodepletion
Quantity 500 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only