missing translation for 'onlineSavingsMsg'
Learn More

IgG, Species Adsorbed: H, R, B, Eq, Gt, Sh, Rb, Ch, Gp Donkey anti-Mouse, AlexaFluor™647, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP192SA6MI
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP192SA6MI 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP192SA6MI Supplier MilliporeSigma Supplier No. AP192SA6

Donkey Polyclonal Antibody

Donkey anti-Mouse IgG, Alexa Fluor 647 Conjugate, Species Adsorbed: H, R, B, Eq, Gt, Sh, Rb, Ch, Gp Antibody is an antibody against Mouse IgG for use in Immunofluorescence.

Specifications

Antigen Donkey anti-Mouse IgG Species adsorbed: H R B Eq Gt Sh Rb Ch Gp
Applications Immunofluorescence
Classification Polyclonal
Concentration 0.5mg
Conjugate Alexa Fluor 647
Formulation Lyophilized in buffer containing 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA and 0.05% sodium azide.
Host Species Donkey
Purification Method Affinity Purified
Quantity 0.5 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C for up to 12 months.
Show More Show Less

For Research Use Only