missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Guinea Pig, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP193PMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP193PMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP193PMI Supplier MilliporeSigma Supplier No. AP193P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Donkey Polyclonal Antibody

Donkey anti-Guinea Pig IgG Antibody, HRP conjugate, Species Adsorbed detects level of Donkey Guinea Pig IgG and has been published and validated for use in ELISA and western blotting.

Specifications

Antigen Donkey anti-Guinea Pig IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate HRP
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA and 0.01% thimerosal. RECONSTITUTION: Reconstitute with 500μL of sterile distilled water.
Host Species Donkey
Quantity 500 μL
Primary or Secondary Secondary
Target Species Guinea Pig
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only