missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Guinea Pig, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP193FMI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP193FMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP193FMI Supplier MilliporeSigma Supplier No. AP193F

Donkey Polyclonal Antibody

This Donkey anti-Guinea Pig IgG Antibody, FITC conjugate, Species Adsorbed is validated for use in IF for the detection of Donkey Guinea Pig IgG.

Specifications

Antigen Donkey anti-Guinea Pig IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Host Species Donkey
Quantity 500 μg
Primary or Secondary Secondary
Target Species Guinea Pig

For Research Use Only