missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Guinea Pig, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP193CMI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP193CMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP193CMI Supplier MilliporeSigma Supplier No. AP193C
Only null left

Donkey Polyclonal Antibody

Detect Donkey Guinea Pig IgG using this Donkey anti-Guinea Pig IgG Antibody, Cy3 conjugate, Species Adsorbed validated for use in immunofluoresence.

Specifications

Antigen Donkey anti-Guinea Pig IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Conjugate Cyanine3
Host Species Donkey
Quantity 500 μg
Primary or Secondary Secondary
Target Species Guinea Pig

For Research Use Only