missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Guinea Pig, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP193BMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP193BMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP193BMI Supplier MilliporeSigma Supplier No. AP193B

Donkey Polyclonal Antibody

Donkey anti-Guinea Pig IgG Antibody, biotin conjugate, Species Adsorbed is an antibody against Donkey Guinea Pig IgG for use in ELISA and western blotting.

Specifications

Antigen Donkey anti-Guinea Pig IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Host Species Donkey
Quantity 500 μL
Primary or Secondary Secondary
Target Species Guinea Pig

For Research Use Only