missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Goat, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP180MI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP180MI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP180MI Supplier MilliporeSigma Supplier No. AP180

Donkey Polyclonal Antibody

Donkey anti-Goat IgG Antibody, Species Adsorbed detects level of Donkey Goat IgG and has been published and validated for use in ELISA, immunoprecipitation and western blotting.

Specifications

Antigen Donkey anti-Goat IgG Species adsorbed
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration ∼1mg/mL
Conjugate Unconjugated
Formulation Liquid in 0.1M Sodium Phosphate, 0.25M NaCl, pH 7.6
Host Species Donkey
Quantity 1 mL
Primary or Secondary Secondary
Target Species Goat
Content And Storage 2-8°C for up to 12 months.
Form Liquid
Show More Show Less

For Research Use Only