missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Donkey anti-Goat IgG (H+L) Secondary Antibody, PE

Donkey Polyclonal Secondary Antibody

Supplier:  Invitrogen™ PA129953

Catalog No. PA129953


Only null left

Description

Description

PA1-29953 detects Goat IgG from goat samples.

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Chemical Identifiers

Goat IgG (H+L)
4°C, store in dark, DO NOT FREEZE!
Polyclonal
Liquid
IgG F(ab')2
Goat
Goat IgG whole molecule.
Secondary Antibody
1.032 mg/mL
Dot Blot, Flow Cytometry, Immunocytochemistry
PE
Donkey
RUO
PBS with 1% BSA and 0.01% sodium azide; pH 7.2
Secondary
Antigen affinity chromatography

Specifications

Goat IgG (H+L)
Polyclonal
PE
Donkey
Antigen affinity chromatography
RUO
Goat
Secondary Antibody
IgG F(ab')2
Dot Blot, Flow Cytometry, Immunocytochemistry
1.032 mg/mL
PBS with 1% BSA and 0.01% sodium azide; pH 7.2
Goat IgG whole molecule.
500 μL
Secondary
4°C, store in dark, DO NOT FREEZE!
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions