missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP

Catalog No. PA186326
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA186326 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA186326 Supplier Invitrogen™ Supplier No. PA186326

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Donkey Polyclonal Secondary Antibody

Store at 4°C prior to reconstitution. After reconstitution store at -20°C. For long term storage the addition of 0.01% thiomersal is recommended. Reconstitute with 0.5 mL of distilled water. Donkey anti goat IgG antibody recognizes goat immunoglobulin G. This antibody has been absorbed against human, rabbit, rat, horse, guinea pig, hamster, chicken and mouse to minimize naturally occurring cross reactivities.

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Goat IgG (H+L) Cross-Adsorbed
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate HRP
Formulation PBS with 1.5% BSA and no preservative
Host Species Donkey
Immunogen Proprietary Immunogen.
Purification Method Affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Goat
Content And Storage 4°C
Product Type Secondary Antibody
Form Lyophilized
Isotype IgG
Show More Show Less