missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Goat, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP180FMI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP180FMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP180FMI Supplier MilliporeSigma Supplier No. AP180F

Donkey Polyclonal Antibody

Detect Donkey Goat IgG using this Donkey anti-Goat IgG Antibody, FITC conjugate, Species Adsorbed validated for use in immunofluoresence.

Specifications

Antigen Donkey anti-Goat IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, and 0.5% sodium azide. RECONSTITUTION: Reconstitute with 500μL of sterile distilled water. FLUOROPHORE/PROTEIN: Approximately 8.2μg/mg.
Host Species Donkey
Quantity 500 μg
Primary or Secondary Secondary
Target Species Goat
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only