missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Donkey anti-Goat, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
This Donkey anti-Goat IgG Antibody, Cy3 conjugate, Species Adsorbed is validated for use in IF for the detection of Donkey Goat IgG.
Specifications
Specifications
| Antigen | Donkey anti-Goat IgG Species adsorbed |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Cyanine3 |
| Host Species | Donkey |
| Quantity | 500 μg |
| Primary or Secondary | Secondary |
| Target Species | Goat |
For Research Use Only