missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Goat, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP180BMI
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP180BMI 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP180BMI Supplier MilliporeSigma Supplier No. AP180B

Donkey Polyclonal Antibody

Donkey anti-Goat IgG Antibody, biotin-SP conjugate, Species Adsorbed detects level of Donkey Goat IgG and has been published and validated for use in ELISA, flow cytometry, immunofluoresence, immunohistochemistry and western blotting.

Specifications

Antigen Donkey anti-Goat IgG Species adsorbed
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Biotin
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, and 0.05% sodium azide. RECONSTITUTION: Reconstitute with 500μL of sterile distilled water.
Host Species Donkey
Purification Method Immunoaffinity Purified and Immunodepletion
Quantity 500 mL
Primary or Secondary Secondary
Target Species Goat
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only