missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Goat, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP180AMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP180AMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP180AMI Supplier MilliporeSigma Supplier No. AP180A

Donkey Polyclonal Antibody

This Donkey anti-Goat IgG Antibody, Alkaline Phosphatase conjugate, Species Adsorbed is validated for use in ELISA, western blotting for the detection of Donkey Goat IgG.

Specifications

Antigen Donkey anti-Goat IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.6mg/mL
Conjugate Alkaline Phosphatase (AP)
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 8.0. RECONSTITUTION: Reconstitute to 1.0mg/mL with 0.5mLsterile distilled water.
Host Species Donkey
Quantity 500 μL
Primary or Secondary Secondary
Target Species Goat
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only