missing translation for 'onlineSavingsMsg'
Learn More

IgY Donkey anti-Chicken, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP194PMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP194PMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP194PMI Supplier MilliporeSigma Supplier No. AP194P
Only null left

Donkey Polyclonal Antibody

This Donkey anti-Chicken IgY Antibody, HRP conjugate, Species Adsorbed is validated for use in ELISA, western blotting for the detection of Donkey Chicken IgY.

Specifications

Antigen Donkey anti-Chicken IgY Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Host Species Donkey
Quantity 500 μL
Primary or Secondary Secondary
Target Species Chicken

For Research Use Only