missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Donkey anti-Chicken IgY (H+L) Secondary Antibody, FITC

Catalog No. SA172000
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA172000 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA172000 Supplier Invitrogen™ Supplier No. SA172000

Donkey Polyclonal Secondary Antibody

SA1-72000 detects Chicken IgY in chicken samples. SA1-72000 has been successfully used in Western Blot, and ELISA procedures. The SA1-72000 immunogen is from purified chicken IgY isolated from normal Chicken eggs.

Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Chicken IgY (H+L)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 50% glycerol and 0.075% sodium azide; pH 7.2
Host Species Donkey
Immunogen Purified chicken IgY isolated from normal chicken eggs.
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Chicken
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less