missing translation for 'onlineSavingsMsg'
Learn More

IgY Donkey anti-Chicken, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP194FMI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP194FMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP194FMI Supplier MilliporeSigma Supplier No. AP194F

Donkey Polyclonal Antibody

Donkey anti-Chicken IgY Antibody, FITC conjugate, Species Adsorbed detects level of Donkey Chicken IgY and has been published and validated for use in immunofluoresence.

Specifications

Antigen Donkey anti-Chicken IgY Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Host Species Donkey
Quantity 500 μg
Primary or Secondary Secondary
Target Species Chicken

For Research Use Only