missing translation for 'onlineSavingsMsg'
Learn More

IgY Donkey anti-Chicken, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP194BMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP194BMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP194BMI Supplier MilliporeSigma Supplier No. AP194B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Donkey Polyclonal Antibody

Donkey anti-Chicken IgY Antibody, biotin conjugate, Species Adsorbed is an antibody against Donkey Chicken IgY for use in ELISA and western blotting.

Specifications

Antigen Donkey anti-Chicken IgY Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Host Species Donkey
Quantity 500 μL
Primary or Secondary Secondary
Target Species Chicken

For Research Use Only