missing translation for 'onlineSavingsMsg'
Learn More

Donepezil hydrochloride

Catalog No. AC458050050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458050050 5 g
AC458050010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458050050 Supplier Thermo Scientific Chemicals Supplier No. 458050050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 120011-70-3 | C24H30ClNO3 | 415.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120011-70-3
Molecular Formula C24H30ClNO3
Molecular Weight (g/mol) 415.96
MDL Number MFCD00881312
InChI Key XWAIAVWHZJNZQQ-UHFFFAOYNA-N
Synonym donepezil hydrochloride, donepezil hcl, aricept, bnag, donepezilhcl, aricept odt, 2-1-benzylpiperidin-4-yl methyl-5,6-dimethoxy-2,3-dihydro-1h-inden-1-one hydrochloride, e 2020 pharmaceutical, eranz
PubChem CID 5741
ChEBI CHEBI:4696
IUPAC Name 2-[(1-benzylpiperidin-4-yl)methyl]-5,6-dimethoxy-2,3-dihydroinden-1-one;hydrochloride
SMILES [H+].[Cl-].COC1=CC2=C(C=C1OC)C(=O)C(CC1CCN(CC3=CC=CC=C3)CC1)C2

Specifications

Melting Point 211.0°C to 212.0°C
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hot methanol, insoluble in ethyl acetate and hexane
Formula Weight 415.85
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Donepezil hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/

RUO – Research Use Only