missing translation for 'onlineSavingsMsg'
Learn More

Donepezil Hydrochloride 98.0+%, TCI America™

Catalog No. p-7128097
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40991G 1 g
D40995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40991G Supplier TCI America Supplier No. D40991G

Chemical Identifiers

CAS 120011-70-3
Molecular Formula C24H30ClNO3
Molecular Weight (g/mol) 415.96
MDL Number MFCD00881312
InChI Key XWAIAVWHZJNZQQ-UHFFFAOYNA-N
Synonym donepezil hydrochloride, donepezil hcl, aricept, bnag, donepezilhcl, aricept odt, 2-1-benzylpiperidin-4-yl methyl-5,6-dimethoxy-2,3-dihydro-1h-inden-1-one hydrochloride, e 2020 pharmaceutical, eranz
PubChem CID 5741
ChEBI CHEBI:4696
IUPAC Name hydrogen 2-[(1-benzylpiperidin-4-yl)methyl]-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one chloride
SMILES [H+].[Cl-].COC1=CC2=C(C=C1OC)C(=O)C(CC1CCN(CC3=CC=CC=C3)CC1)C2

Specifications

Melting Point 225°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 415.96
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Donepezil Hydrochloride
RTECSNumber NK8927885
TSCA No