missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Dog BSEP Vesicles

Catalog No. GM0019
Encompass
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GM0019 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GM0019 Supplier Gibco™ Supplier No. GM0019

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dog BSEP Vesicles are insect derived purified plasma membranes with inserted BSEP transport protein (bile salt export pump, ABCB11).

Our Dog BSEP Vesicles can be used to evaluate test compounds and drug interactions with BSEP in in vitro assays. BSEP is an ATP-binding cassette (ABC) transporter that plays a key role in bile acid excretion from the liver.

Use Dog BSEP Vesicles to:

• Investigate the transporter interactions of your drug candidates
• Assess potential for transporter-mediated drug-drug interactions
• Obtain high quality results with a large signal to noise ratios

Concept Behind ABC Transporter Vesicles
ABC transporter vesicles are easy-to-use, efficient reagents for early assessment of a drug candidate's substrate and drug interaction potential. While ABC transporters typically mediate the export of substrates out of cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for your compound by determining the amount incorporated into the vesicles.

Clear and Reliable Results
Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, these 'inside-out' vesicles provide high levels of transporter activity with low background, giving you a clear signal if your compound is a substrate or inhibitor of a specific efflux transporter.

Transporter Vesicle Details
Dog BSEP Vesicles are part of a larger group of transporter vesicles known as ABC transporter vesicles. They are prepared from purified plasma membranes isolated from an insect cell system (Sf9 cells transfected with baculovirus) expressing BSEP. Evaluation of the inhibitory activity of newly developed compounds towards BSEP-mediated transport of bile acids (e.g. taurocholate) may give an indication of potential hepatotoxicity of the compounds.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Specifications

Quantity 0.5 mL
Purity Research Grade
Host Species Canine
Product Type Dog Vesicle
Content And Storage Store at -80°C.
Format Vial
Shipping Condition Dry Ice
Physical Form Frozen

For Research Use Only. Not for use in diagnostic procedures.