missing translation for 'onlineSavingsMsg'
Learn More

Dodecyltrimethylammonium Bromide 98.0+%, TCI America™

Catalog No. p-7126741
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D146825G 25 g
D1468100G 100 g
D1468500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D146825G Supplier TCI America Supplier No. D146825G

Chemical Identifiers

CAS 1119-94-4
Molecular Formula C15H34BrN
Molecular Weight (g/mol) 308.348
MDL Number MFCD00011767
InChI Key XJWSAJYUBXQQDR-UHFFFAOYSA-M
Synonym dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide
PubChem CID 14249
ChEBI CHEBI:282662
IUPAC Name dodecyl(trimethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Br-]

Specifications

Color White
Quantity 25 g
Formula Weight 308.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dodecyltrimethylammonium Bromide
EINECSNumber (2)-0184&(9)-1971
RTECSNumber BQ3195000
TSCA Yes