missing translation for 'onlineSavingsMsg'
Learn More

Dodecyltrimethylammonium bromide, 99%, pure

Catalog No. p-4258988
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC409310250 25 g
AC409311000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC409310250 Supplier Thermo Scientific Chemicals Supplier No. 409310250
Only null left

CAS: 1119-94-4 | C15H34BrN | 308.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 1119-94-4
Molecular Formula C15H34BrN
Molecular Weight (g/mol) 308.34
InChI Key XJWSAJYUBXQQDR-UHFFFAOYSA-M
Synonym dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide
PubChem CID 14249
ChEBI CHEBI:282662
IUPAC Name dodecyl(trimethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Br-]

Specifications

Melting Point 246.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3(CH2)11N(Br)(CH3)3
Quantity 25 g
Solubility Information (10% in water) Clear to slightly hazy
Formula Weight 308.34
Percent Purity 99%
Grade Pure
Physical Form Powder
Chemical Name or Material Dodecyltrimethylammonium bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
EINECSNumber 214-290-3

RUO – Research Use Only