missing translation for 'onlineSavingsMsg'
Learn More

Dodecyltrimethoxysilane 93.0+%, TCI America™

Catalog No. p-7127705
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D338325G 25 g
D3383250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D338325G Supplier TCI America Supplier No. D338325G
Only null left

Chemical Identifiers

CAS 3069-21-4
Molecular Formula C15H34O3Si
Molecular Weight (g/mol) 290.52
MDL Number MFCD00069148
InChI Key SCPWMSBAGXEGPW-UHFFFAOYSA-N
Synonym n-dodecyltrimethoxysilane, silane, dodecyltrimethoxy, lauryltrimethoxysilane, dodecyl trimethoxy silane, n-dodecytrimethoxysilane, acmc-1cr18, ksc494k8f
PubChem CID 76479
IUPAC Name dodecyltrimethoxysilane
SMILES CCCCCCCCCCCC[Si](OC)(OC)OC

Specifications

Melting Point -40°C
Color Colorless
Boiling Point 118°C
Quantity 25 g
Formula Weight 290.52
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecyltrimethoxysilane