accessibility menu, dialog, popup

UserName

H3CSiOCH3OCH3OCH3

CAS RN 3069-21-4

IUPAC Name: dodecyltrimethoxysilane
Synonyms: dodecyltrimethoxysilane
Molecular Weight (g/mol): 290.52
Molecular Formula: C15H34O3Si
InChi Key: SCPWMSBAGXEGPW-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC[Si](OC)(OC)OC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results