missing translation for 'onlineSavingsMsg'
Learn More

Dodecylphosphonic Acid 98.0+%, TCI America™

Catalog No. p-7128476
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48091G 1 g
D48095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48091G Supplier TCI America Supplier No. D48091G
Only null left

Chemical Identifiers

CAS 5137-70-2
Molecular Formula C12H27O3P
Molecular Weight (g/mol) 250.32
MDL Number MFCD00015832
InChI Key SVMUEEINWGBIPD-UHFFFAOYSA-N
Synonym n-dodecylphosphonic acid, phosphonic acid, dodecyl, n-dodecanephosphonic acid, dodecane phosphonic acid, 1-dodecanephosphonic acid, n-dodecyl phosphonic acid, dodecyl-phosphonic acid, 1-dodecylphosphonic acid, phosphonic acid, p-dodecyl, n-dodecylphosphonicacid
PubChem CID 78816
IUPAC Name dodecylphosphonic acid
SMILES CCCCCCCCCCCCP(O)(O)=O

Specifications

Melting Point 101°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 250.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dodecylphosphonic Acid
RTECSNumber SZ7875000
TSCA Yes