missing translation for 'onlineSavingsMsg'
Learn More

Dodecylcyclohexane 98.0+%, TCI America™

Catalog No. p-7126642
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D123225ML 25 mL
D1232500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D123225ML Supplier TCI America Supplier No. D123225ML

Chemical Identifiers

CAS 1795-17-1
Molecular Formula C18H36
Molecular Weight (g/mol) 252.486
MDL Number MFCD00045506
InChI Key BLRBGKYYWDBAQQ-UHFFFAOYSA-N
Synonym 1-Cyclohexyldodecane, Laurylcyclohexane
PubChem CID 15714
IUPAC Name dodecylcyclohexane
SMILES CCCCCCCCCCCCC1CCCCC1

Specifications

Color Colorless
Boiling Point 126°C
Quantity 25 mL
Formula Weight 252.49
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecylcyclohexane