accessibility menu, dialog, popup

UserName

CAS RN 1795-17-1

H3C

CAS RN 1795-17-1

IUPAC Name: dodecylcyclohexane
Synonyms: dodecylcyclohexane
Molecular Weight (g/mol): 252.49
Molecular Formula: C18H36
InChi Key: BLRBGKYYWDBAQQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results