missing translation for 'onlineSavingsMsg'
Learn More

Dodecylamine Acetate 98.0+%, TCI America™

Catalog No. D098125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D098125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D098125G Supplier TCI America Supplier No. D098125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2016-56-0
Molecular Formula C14H31NO2
Molecular Weight (g/mol) 245.41
MDL Number MFCD00043524
InChI Key HBRNMIYLJIXXEE-UHFFFAOYSA-N
Synonym 1-Aminododecane Acetate, Laurylamine Acetate, Dodecan-1-amine Acetate
PubChem CID 263045
IUPAC Name acetic acid; dodecan-1-amine
SMILES CC(O)=O.CCCCCCCCCCCCN

Specifications

Melting Point 69°C
Color White
Quantity 25 g
Formula Weight 245.41
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dodecylamine Acetate
RTECSNumber JR6490000
TSCA Yes