accessibility menu, dialog, popup

UserName

CAS RN 2016-56-0

IUPAC Name: acetic acid; dodecan-1-amine
Synonyms: N-dodecylamine; acetic acid
Molecular Weight (g/mol): 245.41
Molecular Formula: C14H31NO2
InChi Key: HBRNMIYLJIXXEE-UHFFFAOYSA-N
SMILES: CC(O)=O.CCCCCCCCCCCCN
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results