missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

Supplier:  Thermo Scientific™ 419530250

 View more versions of this product

Catalog No. AC419530250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry
Specifications

Specifications

Dodecyl sulfate, sodium salt
151-21-3,7757-82-6,7647-14-5
Fine Crystalline, Flaky or Granular Powder
98%
CH3(CH2)11OSO3Na
01, III, 1786
>150°C
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
3423265
288.38
Biochemical
1% max.
206.0°C
White
25 g
C12H25NaO4S
MFCD00036175
13,281
15, 8768
Solubility in water: 130g/L (20°C). Other solubilities: partly soluble in alcohol, soluble 75g/L in ethanol (20°C)
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
98%
Authentic
3% aq. soln. clear, colorless. 1% soln. in EtOH clear, colorless
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only