missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis

Catalog No. p-4258712
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC419530250 25 g
AC419531000 100 g
AC419530010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC419530250 Supplier Thermo Scientific Chemicals Supplier No. 419530250
Only null left

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry

Specifications

Chemical Name or Material Dodecyl sulfate, sodium salt
Melting Point 206.0°C
Color White
CAS 151-21-3, 7757-82-6, 7647-14-5
Physical Form Fine Crystalline, Flaky or Granular Powder
Quantity 25 g
Assay Percent Range 98%
Molecular Formula C12H25NaO4S
Linear Formula CH3(CH2)11OSO3Na
MDL Number MFCD00036175
Beilstein 01, III, 1786
Flash Point >150°C
Fieser 13,281
Merck Index 15, 8768
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
Solubility Information Solubility in water: 130g/L (20°C). Other solubilities: partly soluble in alcohol, soluble 75g/L in ethanol (20°C)
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
IUPAC Name sodium dodecyl sulfate
Molecular Weight (g/mol) 288.38
PubChem CID 3423265
ChEBI CHEBI:8984
Formula Weight 288.38
Percent Purity 98%
Grade Biochemical
Infrared Spectrum Authentic
Loss on Drying 1% max.
Appearance 3% aq. soln. clear, colorless. 1% soln. in EtOH clear, colorless
Show More Show Less

RUO – Research Use Only