missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Stearate 60.0+%, TCI America™

Catalog No. S007825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S007825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S007825G Supplier TCI America Supplier No. S007825G
Only null left

Chemical Identifiers

CAS 5303-25-3
Molecular Formula C30H60O2
Molecular Weight (g/mol) 452.808
MDL Number MFCD00059290
InChI Key JRTVEUGOGWTHTR-UHFFFAOYSA-N
Synonym Stearic Acid Dodecyl Ester, Lauryl Stearate, Stearic Acid Lauryl Ester
PubChem CID 79186
IUPAC Name dodecyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCCCCCCCCCCC

Specifications

Melting Point 41°C
Color White
Quantity 25 g
Formula Weight 452.81
Percent Purity ≥60.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dodecyl Stearate
EINECSNumber (2)-0824
TSCA Yes