accessibility menu, dialog, popup

UserName

CAS RN 5303-25-3

H3CONSNOO(E)

CAS RN 5303-25-3

IUPAC Name: (2E)-3-(2-ethoxyphenyl)-2-(4-{3-oxo-3H-benzo[f]chromen-2-yl}-1,3-thiazol-2-yl)prop-2-enenitrile
Synonyms: lauryl stearate
Molecular Weight (g/mol): 450.51
Molecular Formula: C27H18N2O3S
InChi Key: ZUFLXGKRKMOPFG-CPNJWEJPSA-N
SMILES: CCOC1=CC=CC=C1\C=C(/C#N)C1=NC(=CS1)C1=CC2=C(OC1=O)C=CC1=CC=CC=C21
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results