missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl hydrogen sulfate sodium salt, MilliporeSigma™

Catalog No. M1183090025
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1183090025 25 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1183090025 Supplier Technidata Supplier No. 1.18309.0025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For ion pair chromatography LiChropur™

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Melting Point 204.06°C to 207.06°C
Density 1.1g/cm3 (20°C)
pH 6 to 9
Flash Point >150°C (>302°F)
Packaging Plastic Bottle
Quantity 25 g
Formula Weight 288.37
Chemical Name or Material Dodecyl hydrogen sulfate sodium salt
Hazard Category H228: Flammable solid.
Hazard Statement H302: Harmful if swallowed.
Precautionary Statement H315: Causes skin irritation.
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 205-788-1
Recommended Storage 15° to 25°C (59° to 77°F); Tightly Closed. Dry. Keep in well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons.