missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Chloroformate 90.0+%, TCI America™

Catalog No. F022325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F022325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F022325G Supplier TCI America Supplier No. F022325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24460-74-0
Molecular Formula C13H25ClO2
Molecular Weight (g/mol) 248.791
InChI Key AFPOMDNRTZLRMD-UHFFFAOYSA-N
Synonym Chloroformic Acid Dodecyl Ester, Chloroformic Acid Lauryl Ester, Lauryl Chloroformate
PubChem CID 90513
IUPAC Name dodecyl carbonochloridate
SMILES CCCCCCCCCCCCOC(=O)Cl

Specifications

Color Yellow
Boiling Point 168°C
Quantity 25 g
UN Number 3277
Formula Weight 248.79
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecyl Chloroformate