accessibility menu, dialog, popup

UserName

CAS RN 24460-74-0

H3COClO

CAS RN 24460-74-0

IUPAC Name: dodecyl carbonochloridate
Synonyms: dodecyl carbonochloridate
Molecular Weight (g/mol): 248.79
Molecular Formula: C13H25ClO2
InChi Key: AFPOMDNRTZLRMD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results