missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecyl 4-Hydroxybenzoate 98.0+%, TCI America™
$175.22 - $1731.71
Chemical Identifiers
| CAS | 2664-60-0 |
|---|---|
| Molecular Formula | C19H30O3 |
| Molecular Weight (g/mol) | 306.446 |
| MDL Number | MFCD00016473 |
| InChI Key | BAYSQTBAJQRACX-UHFFFAOYSA-N |
| Synonym | dodecyl p-hydroxybenzoate, dodecylparaben, n-dodecyl 4-hydroxybenzoate, p-hydroxybenzoic acid dodecyl ester, benzoic acid, 4-hydroxy-, dodecyl ester, p-oxybenzoesaureduodecylester, benzoic acid, p-hydroxy-, dodecyl ester, p-oxybenzoesaureduodecylester german, 4-hydroxybenzoic acid dodecyl ester, laurylparaben |
| PubChem CID | 17569 |
| IUPAC Name | dodecyl 4-hydroxybenzoate |
| SMILES | CCCCCCCCCCCCOC(=O)C1=CC=C(C=C1)O |
Chemical Identifiers
| 2664-60-0 | |
| 306.446 | |
| BAYSQTBAJQRACX-UHFFFAOYSA-N | |
| 17569 | |
| CCCCCCCCCCCCOC(=O)C1=CC=C(C=C1)O |
| C19H30O3 | |
| MFCD00016473 | |
| dodecyl p-hydroxybenzoate, dodecylparaben, n-dodecyl 4-hydroxybenzoate, p-hydroxybenzoic acid dodecyl ester, benzoic acid, 4-hydroxy-, dodecyl ester, p-oxybenzoesaureduodecylester, benzoic acid, p-hydroxy-, dodecyl ester, p-oxybenzoesaureduodecylester german, 4-hydroxybenzoic acid dodecyl ester, laurylparaben | |
| dodecyl 4-hydroxybenzoate |
Specifications
| 2664-60-0 | |
| White | |
| MFCD00016473 | |
| dodecyl p-hydroxybenzoate, dodecylparaben, n-dodecyl 4-hydroxybenzoate, p-hydroxybenzoic acid dodecyl ester, benzoic acid, 4-hydroxy-, dodecyl ester, p-oxybenzoesaureduodecylester, benzoic acid, p-hydroxy-, dodecyl ester, p-oxybenzoesaureduodecylester german, 4-hydroxybenzoic acid dodecyl ester, laurylparaben | |
| CCCCCCCCCCCCOC(=O)C1=CC=C(C=C1)O | |
| 306.446 | |
| 306.45 | |
| Crystalline Powder |
| 55°C | |
| C19H30O3 | |
| 25 g | |
| BAYSQTBAJQRACX-UHFFFAOYSA-N | |
| dodecyl 4-hydroxybenzoate | |
| 17569 | |
| ≥98.0% (HPLC,T) | |
| Dodecyl 4-Hydroxybenzoate |
Safety and Handling
EINECSNumber : (9)-1074
RTECSNumber : DH2185600
TSCA : No