missing translation for 'onlineSavingsMsg'
Learn More

Dodecanoic acid, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013108
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013108 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013108 Supplier Merck Emd Millipore Supplier No. 616095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 143-07-7
Flash Point 160°C to 165°C
Physical Form Neat
Linear Formula CH3(CH2)10 COOH
MDL Number MFCD00002736
Melting Point 43°C to 46°C
Synonym Lauric acid; NSC 5026
Type Analytical Standard
Quantity 5 g
Formula Weight 200.32
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less