missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecanal (stabilized with DL-alpha-Tocopherol) 95.0+%, TCI America™
$119.36 - $392.74
Chemical Identifiers
| CAS | 112-54-9 |
|---|---|
| Molecular Formula | C12H24O |
| Molecular Weight (g/mol) | 184.32 |
| MDL Number | MFCD00007017 |
| InChI Key | HFJRKMMYBMWEAD-UHFFFAOYSA-N |
| Synonym | lauraldehyde, dodecyl aldehyde, lauric aldehyde, lauryl aldehyde, n-dodecanal, dodecanaldehyde, 1-dodecanal, n-dodecyl aldehyde, n-lauraldehyde, n-dodecylic aldehyde |
| PubChem CID | 8194 |
| ChEBI | CHEBI:27836 |
| IUPAC Name | dodecanal |
| SMILES | CCCCCCCCCCCC=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D097925ML
|
TCI America
D097925ML |
25 mL |
Each for $119.36
|
|
|||||
|
D0979500ML
|
TCI America
D0979500ML |
500 mL |
Each for $392.74
|
|
|||||
Chemical Identifiers
| 112-54-9 | |
| 184.32 | |
| HFJRKMMYBMWEAD-UHFFFAOYSA-N | |
| 8194 | |
| dodecanal |
| C12H24O | |
| MFCD00007017 | |
| lauraldehyde, dodecyl aldehyde, lauric aldehyde, lauryl aldehyde, n-dodecanal, dodecanaldehyde, 1-dodecanal, n-dodecyl aldehyde, n-lauraldehyde, n-dodecylic aldehyde | |
| CHEBI:27836 | |
| CCCCCCCCCCCC=O |
Specifications
| 112-54-9 | |
| 241°C | |
| MFCD00007017 | |
| lauraldehyde, dodecyl aldehyde, lauric aldehyde, lauryl aldehyde, n-dodecanal, dodecanaldehyde, 1-dodecanal, n-dodecyl aldehyde, n-lauraldehyde, n-dodecylic aldehyde | |
| CCCCCCCCCCCC=O | |
| 184.32 | |
| CHEBI:27836 | |
| ≥95.0% (GC) | |
| Dodecanal (stabilized with DL-alpha-Tocopherol) |
| Yellow | |
| C12H24O | |
| 25 mL | |
| HFJRKMMYBMWEAD-UHFFFAOYSA-N | |
| dodecanal | |
| 8194 | |
| 184.32 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0494
RTECSNumber : JR1910000
TSCA : Yes