missing translation for 'onlineSavingsMsg'
Learn More

Dodecahydrofluorene (mixture of isomers) 97.0+%, TCI America™

Catalog No. p23931g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P23931G Supplier TCI America Supplier No. P23931G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5744-03-6
Molecular Formula C13H22
Molecular Weight (g/mol) 178.32
MDL Number MFCD00001161
InChI Key OLWAZOBRCQWWDB-UHFFFAOYNA-N
Synonym Perhydrofluorene
PubChem CID 21972
IUPAC Name 2,3,4,4a,4b,5,6,7,8,8a,9,9a-dodecahydro-1H-fluorene
SMILES C1CCC2C(C1)CC3C2CCCC3

Specifications

Melting Point 15°C
Color Yellow
Boiling Point 253°C
Quantity 1 g
Formula Weight 178.32
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecahydrofluorene (mixture of isomers)