accessibility menu, dialog, popup

UserName

CAS RN 5744-03-6

IUPAC Name: dodecahydro-1H-fluorene
Synonyms: dodecahydrofluorene
Molecular Weight (g/mol): 178.32
Molecular Formula: C13H22
InChi Key: OLWAZOBRCQWWDB-UHFFFAOYNA-N
SMILES: C1C2CCCCC2C2CCCCC12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results