Learn More
Dodecacarbonyltriruthenium, 99%
Description
A H-transfer catalyst. It is used in carbonylation catalysis of the allylic amination of unactivated olefins by nitroarenes, reductive carbonylation of mononitro aromatic compounds to carbamates, and as a catalyst for cycloaddition reactions. It is employed as a precursor for the synthesis of Ru nanoparticles. Catalyst used in the intermolecular [3+2+1] cycloaddition of alpha,beta-unsaturated ketones with silylacetylenes and carbon monoxide, to produce alpha-pyrones. Catalyzes the coupling of silanes with thiols, alcohols and amines with high turnover number (TON) and high turnover frequency (TOF).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼150°C (decomposition) |
| Color | Orange |
| Linear Formula | Ru3(CO)12 |
| Quantity | 1 g |
| UN Number | UN3466 |
| Solubility Information | Sparingly soluble in hydrocarbons (e;g; hexane,cyclohexane,benzene) and acetone. |
| Formula Weight | 639.33 |
| Percent Purity | 99% |
| Physical Form | Crystalline |
| Chemical Name or Material | Dodecacarbonyltriruthenium |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H331 Toxic if inhaled. |
| Precautionary Statement | GHS P Statement P261-P304+P340-P311-P405-P403+P233-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. Store locked up. Store in a well-ventilated place. Keep container tightly closed. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S. |
| EINECSNumber | 239-287-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only