missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doctor Solution (Sodium Plumbite), Ricca Chemical
Supplier: Ricca Chemical Company 26505HP
Specifications
| Doctor Solution | |
| Sodium Plumbite | |
| 86.0,10.71,1.67 | |
| dihydrogen oxide, dihydrogen monoxide | |
| Hard Pack | |
| 20 L |
| Orange | |
| 7732-18-5,1310-73-2,1317-36-8 | |
| 89.0,11.15,1.74 | |
| Laboratory | |
| >12 |