missing translation for 'onlineSavingsMsg'
Learn More

Doctor Solution (Sodium Plumbite) ASTM D 4952, Reagents

Catalog No. 01335431
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335431 4 L
01335432 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335431 Supplier Labchem Inc Supplier No. CD1001004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Doctor Solution (Sodium Plumbite) ASTM D 4952, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Doctor Solution (Sodium Plumbite) ASTM D 4952
Color Colorless with Slight Orange
CAS 7732-18-5, 1310-73-2
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.14
Quantity 4 L
Sterility Non-sterile
Physical Form Liquid