missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Docosyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™
Supplier: TCI America D507525G
Specifications
| Docosyl Acrylate (stabilized with MEHQ) | |
| 47°C | |
| C25H48O2 | |
| 25 g | |
| KHAYCTOSKLIHEP-UHFFFAOYSA-N | |
| docosyl prop-2-enoate | |
| 87565 | |
| ≥95.0% (GC) |
| 18299-85-9 | |
| White-Yellow | |
| MFCD00080409 | |
| Acrylic Acid Docosyl Ester, Behenyl Acrylate, Acrylic Acid Behenyl Ester | |
| CCCCCCCCCCCCCCCCCCCCCCOC(=O)C=C | |
| 380.657 | |
| 380.66 | |
| Crystalline Lumps |
Chemical Identifiers
| 18299-85-9 | |
| 380.657 | |
| KHAYCTOSKLIHEP-UHFFFAOYSA-N | |
| 87565 | |
| CCCCCCCCCCCCCCCCCCCCCCOC(=O)C=C |
| C25H48O2 | |
| MFCD00080409 | |
| Acrylic Acid Docosyl Ester, Behenyl Acrylate, Acrylic Acid Behenyl Ester | |
| docosyl prop-2-enoate |
Safety and Handling
EINECSNumber : (2)-3948
TSCA : Yes