missing translation for 'onlineSavingsMsg'
Learn More

Docosyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. p-7128644
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D507525G 25 g
D5075100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D507525G Supplier TCI America Supplier No. D507525G

Chemical Identifiers

CAS 18299-85-9
Molecular Formula C25H48O2
Molecular Weight (g/mol) 380.657
MDL Number MFCD00080409
InChI Key KHAYCTOSKLIHEP-UHFFFAOYSA-N
Synonym Acrylic Acid Docosyl Ester, Behenyl Acrylate, Acrylic Acid Behenyl Ester
PubChem CID 87565
IUPAC Name docosyl prop-2-enoate
SMILES CCCCCCCCCCCCCCCCCCCCCCOC(=O)C=C

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 25 g
Formula Weight 380.66
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Docosyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-3948
TSCA Yes