missing translation for 'onlineSavingsMsg'
Learn More

Docosane 98.0+%, TCI America™

Catalog No. D096225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D096225G 25 g
D0962100G 100 g
D0962500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D096225G Supplier TCI America Supplier No. D096225G

Chemical Identifiers

CAS 629-97-0
Molecular Formula C22H46
Molecular Weight (g/mol) 310.61
MDL Number MFCD00009348
InChI Key HOWGUJZVBDQJKV-UHFFFAOYSA-N
Synonym n-docosane, dokosan, unii-ow99q363ko, heneicosane, methyl, ch3-ch2 20-ch3, docosane, analytical standard, twt, normal-docosane, docosane, n, docosane
PubChem CID 12405
ChEBI CHEBI:46050
IUPAC Name docosane
SMILES CCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 46°C
Color White
Boiling Point 380°C
Quantity 25 g
Formula Weight 310.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Docosane
EINECSNumber (2)-0010
TSCA Yes